Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGTTGAATGGGGGTATGGAAGC[C/T]ACCTTAGGCAAGATTTCCAGAGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
EML6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EML6 - echinoderm microtubule associated protein like 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039753.2 | Intron | NP_001034842.2 | ||||
XM_017004098.1 | Intron | XP_016859587.1 | ||||
XM_017004099.1 | Intron | XP_016859588.1 | ||||
XM_017004100.1 | Intron | XP_016859589.1 | ||||
XM_017004101.1 | Intron | XP_016859590.1 | ||||
XM_017004102.1 | Intron | XP_016859591.1 |