Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATAAATGGGCTCTCAGGATTCTT[A/T]GGGTCAATATTAGTGAAAATAAACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607496 MIM: 609395 | ||||||||||||||||||||
Literature Links: |
NOSTRIN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NOSTRIN - nitric oxide synthase trafficking | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPC25 - SPC25, NDC80 kinetochore complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020675.3 | 629 | Silent Mutation | CCA,CCT | P,P 163 | NP_065726.1 | |
XM_011511516.2 | 629 | Silent Mutation | CCA,CCT | P,P 163 | XP_011509818.1 |