Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGTGGGACCATGCCAGGCACAAAA[C/T]GGTTTCAACATGTCATTGAGACCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611905 MIM: 600842 MIM: 607386 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FNDC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FNDC4 - fibronectin type III domain containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GCKR - glucokinase (hexokinase 4) regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001486.3 | 82 | Missense Mutation | CGG,TGG | R,W 6 | NP_001477.2 | |
XM_011532763.1 | 82 | Missense Mutation | CGG,TGG | R,W 6 | XP_011531065.1 | |
XM_017003796.1 | 82 | Intron | XP_016859285.1 | |||
XM_017003797.1 | 82 | Intron | XP_016859286.1 |
IFT172 - intraflagellar transport 172 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |