Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAATAGACTCACTCTTCTAATGAT[A/C]GATCTAGGCCTCGGTCAGGAGATCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607166 | ||||||||||||||||||||
Literature Links: |
TSGA10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TSGA10 - testis specific 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025244.2 | 2442 | Missense Mutation | CGA,CTA | R,L 687 | NP_079520.1 | |
NM_182911.3 | 2442 | Missense Mutation | CGA,CTA | R,L 687 | NP_878915.2 | |
XM_005264023.1 | 2442 | Missense Mutation | CGA,CTA | R,L 687 | XP_005264080.1 | |
XM_005264024.2 | 2442 | Missense Mutation | CGA,CTA | R,L 687 | XP_005264081.1 | |
XM_005264025.1 | 2442 | Missense Mutation | CGA,CTA | R,L 687 | XP_005264082.1 | |
XM_005264026.1 | 2442 | Missense Mutation | CGA,CTA | R,L 687 | XP_005264083.1 | |
XM_005264027.1 | 2442 | Missense Mutation | CGA,CTA | R,L 687 | XP_005264084.1 | |
XM_005264029.1 | 2442 | Missense Mutation | CGA,CTA | R,L 687 | XP_005264086.1 | |
XM_006712781.2 | 2442 | Missense Mutation | CGA,CTA | R,L 687 | XP_006712844.1 | |
XM_011511934.1 | 2442 | Missense Mutation | CGA,CTA | R,L 887 | XP_011510236.1 | |
XM_011511935.2 | 2442 | Missense Mutation | CGA,CTA | R,L 815 | XP_011510237.1 | |
XM_011511937.1 | 2442 | Missense Mutation | CGA,CTA | R,L 687 | XP_011510239.1 | |
XM_011511939.1 | 2442 | Missense Mutation | CGA,CTA | R,L 687 | XP_011510241.1 | |
XM_011511940.2 | 2442 | Missense Mutation | CGA,CTA | R,L 687 | XP_011510242.1 | |
XM_011511941.1 | 2442 | Missense Mutation | CGA,CTA | R,L 687 | XP_011510243.1 | |
XM_011511942.1 | 2442 | Missense Mutation | CGA,CTA | R,L 687 | XP_011510244.1 | |
XM_011511943.2 | 2442 | Missense Mutation | CGA,CTA | R,L 687 | XP_011510245.1 | |
XM_017005035.1 | 2442 | Missense Mutation | CGA,CTA | R,L 882 | XP_016860524.1 | |
XM_017005036.1 | 2442 | Intron | XP_016860525.1 | |||
XM_017005037.1 | 2442 | Intron | XP_016860526.1 | |||
XM_017005038.1 | 2442 | Intron | XP_016860527.1 | |||
XM_017005039.1 | 2442 | Missense Mutation | CGA,CTA | R,L 687 | XP_016860528.1 | |
XM_017005040.1 | 2442 | Missense Mutation | CGA,CTA | R,L 687 | XP_016860529.1 | |
XM_017005041.1 | 2442 | Missense Mutation | CGA,CTA | R,L 687 | XP_016860530.1 | |
XM_017005042.1 | 2442 | Missense Mutation | CGA,CTA | R,L 687 | XP_016860531.1 | |
XM_017005043.1 | 2442 | Missense Mutation | CGA,CTA | R,L 687 | XP_016860532.1 | |
XM_017005044.1 | 2442 | Missense Mutation | CGA,CTA | R,L 687 | XP_016860533.1 | |
XM_017005045.1 | 2442 | Missense Mutation | CGA,CTA | R,L 687 | XP_016860534.1 | |
XM_017005046.1 | 2442 | Missense Mutation | CGA,CTA | R,L 687 | XP_016860535.1 | |
XM_017005047.1 | 2442 | Missense Mutation | CGA,CTA | R,L 687 | XP_016860536.1 | |
XM_017005048.1 | 2442 | Missense Mutation | CGA,CTA | R,L 687 | XP_016860537.1 |