Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGTAGAAGTCTTTTATAGTTAGC[A/T]GAGGTGGAACAAGTGGGTCAGCAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 157655 | ||||||||||||||||||||
Literature Links: |
GCSHP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GCSHP3 - glycine cleavage system protein H pseudogene 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFS1 - NADH:ubiquinone oxidoreductase core subunit S1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199981.1 | 2166 | Missense Mutation | CAG,CTG | Q,L 652 | NP_001186910.1 | |
NM_001199982.1 | 2166 | Missense Mutation | CAG,CTG | Q,L 577 | NP_001186911.1 | |
NM_001199983.1 | 2166 | Missense Mutation | CAG,CTG | Q,L 631 | NP_001186912.1 | |
NM_001199984.1 | 2166 | Missense Mutation | CAG,CTG | Q,L 702 | NP_001186913.1 | |
NM_005006.6 | 2166 | Missense Mutation | CAG,CTG | Q,L 688 | NP_004997.4 | |
XM_017004188.1 | 2166 | Missense Mutation | CAG,CTG | Q,L 435 | XP_016859677.1 |