Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCTAGCCCACACCAGTTATCCT[A/G]TTGAAAGAGGGGACTGATAGCTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605140 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCT7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCT7 - chaperonin containing TCP1 subunit 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009570.2 | 167 | Intron | NP_001009570.1 | |||
NM_001166284.1 | 167 | Intron | NP_001159756.1 | |||
NM_001166285.1 | 167 | UTR 5 | NP_001159757.1 | |||
NM_006429.3 | 167 | Silent Mutation | CTA,CTG | L,L 8 | NP_006420.1 | |
XM_011532478.2 | 167 | UTR 5 | XP_011530780.1 | |||
XM_011532479.1 | 167 | UTR 5 | XP_011530781.1 |
PRADC1 - protease associated domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |