Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCAGTGAGGACGGGACTCTACTGC[C/T]GAGACCAGGCTCACGCTGAGAGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605096 MIM: 602877 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANO7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANO7 - anoctamin 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001666.3 | 149 | Nonsense Mutation | CGA,TGA | R,* 30 | NP_001001666.1 | |
NM_001001891.3 | 149 | Nonsense Mutation | CGA,TGA | R,* 30 | NP_001001891.2 | |
XM_011511263.2 | 149 | Nonsense Mutation | CGA,TGA | R,* 30 | XP_011509565.1 | |
XM_011511267.2 | 149 | Nonsense Mutation | CGA,TGA | R,* 30 | XP_011509569.1 | |
XM_017004229.1 | 149 | Nonsense Mutation | CGA,TGA | R,* 30 | XP_016859718.1 | |
XM_017004230.1 | 149 | Intron | XP_016859719.1 |
PPP1R7 - protein phosphatase 1 regulatory subunit 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |