Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCCAACATCGTGGACATGCTCTA[C/T]CTCAAGGACTTGGCCTTCGTGGATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607804 MIM: 607805 | ||||||||||||||||||||
Literature Links: |
CNNM3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CNNM3 - cyclin and CBS domain divalent metal cation transport mediator 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017623.4 | 1116 | Silent Mutation | TAC,TAT | Y,Y 364 | NP_060093.3 | |
NM_199078.2 | 1116 | Silent Mutation | TAC,TAT | Y,Y 364 | NP_951060.1 | |
XM_011510957.2 | 1116 | Silent Mutation | TAC,TAT | Y,Y 364 | XP_011509259.1 | |
XM_017003800.1 | 1116 | Silent Mutation | TAC,TAT | Y,Y 364 | XP_016859289.1 |
CNNM4 - cyclin and CBS domain divalent metal cation transport mediator 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |