Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGCCCTCTCAGCAAGAACGGGAA[C/T]AAAACTCTGGTAATCGCTTCTGTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612282 | ||||||||||||||||||||
Literature Links: |
LOC105373780 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC105373780 - proline-rich protein 18-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011512285.1 | 696 | Intron | XP_011510587.1 |
ZNF804A - zinc finger protein 804A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_194250.1 | 696 | Silent Mutation | AAC,AAT | N,N 34 | NP_919226.1 |