Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCACACACTTTATGTTTTGTAAAT[G/T]TTCTGTAATCCTTTTGCACACAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611148 MIM: 604277 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC30A6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC30A6 - solute carrier family 30 member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_006712048.3 | Intron | XP_006712111.1 | ||||
XM_011532959.2 | Intron | XP_011531261.1 | ||||
XM_011532960.2 | Intron | XP_011531262.1 | ||||
XM_011532961.2 | Intron | XP_011531263.1 | ||||
XM_011532962.2 | Intron | XP_011531264.1 | ||||
XM_011532963.2 | Intron | XP_011531265.1 | ||||
XM_017004462.1 | Intron | XP_016859951.1 | ||||
XM_017004463.1 | Intron | XP_016859952.1 | ||||
XM_017004464.1 | Intron | XP_016859953.1 | ||||
XM_017004465.1 | Intron | XP_016859954.1 | ||||
XM_017004466.1 | Intron | XP_016859955.1 |
SPAST - spastin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |