Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGACCCTTGAGAAAGACAGAGG[A/G]AACCCCATCTACCTGGGCCTGAATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605509 | ||||||||||||||||||||
Literature Links: |
IL36A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IL36A - interleukin 36, alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014440.2 | 591 | Silent Mutation | GGA,GGG | G,G 60 | NP_055255.1 | |
XM_005263639.2 | 591 | Silent Mutation | GGA,GGG | G,G 60 | XP_005263696.1 | |
XM_011510965.1 | 591 | Silent Mutation | GGA,GGG | G,G 60 | XP_011509267.1 | |
XM_017003806.1 | 591 | Silent Mutation | GGA,GGG | G,G 60 | XP_016859295.1 |