Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCTCCAGTCCTCTGCCGTAGTGG[C/T]CAGAGCTCTGAGCGGGGGCGCGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603301 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYS1 - cystin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KLF11 - Kruppel like factor 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177716.1 | 454 | Missense Mutation | GCC,GTC | A,V 129 | NP_001171187.1 | |
NM_001177718.1 | 454 | Missense Mutation | GCC,GTC | A,V 129 | NP_001171189.1 | |
NM_003597.4 | 454 | Missense Mutation | GCC,GTC | A,V 146 | NP_003588.1 |
LOC101929882 - uncharacterized LOC101929882 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |