Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACTACACCCTTGCACCCCCAGAA[C/T]GAGCGAGAGGAAGCCCTCACCACCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 100720 MIM: 100730 MIM: 612972 | ||||||||||||||||||||
Literature Links: |
CHRND PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHRND - cholinergic receptor nicotinic delta subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CHRNG - cholinergic receptor nicotinic gamma subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005199.4 | 219 | Silent Mutation | AAC,AAT | N,N 66 | NP_005190.4 |
TIGD1 - tigger transposable element derived 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |