Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGACCCCATCTTCGTCCGCAACC[A/T]CCTCGAACGCGGGAGCGGACCCCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605363 | ||||||||||||||||||||
Literature Links: |
GAD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GAD1 - glutamate decarboxylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000817.2 | 454 | Missense Mutation | ACC,TCC | T,S 12 | NP_000808.2 | |
NM_013445.3 | 454 | Missense Mutation | ACC,TCC | T,S 12 | NP_038473.2 | |
XM_005246444.2 | 454 | Missense Mutation | ACC,TCC | T,S 12 | XP_005246501.1 | |
XM_011510922.1 | 454 | Missense Mutation | ACC,TCC | T,S 12 | XP_011509224.1 | |
XM_017003756.1 | 454 | Missense Mutation | ACC,TCC | T,S 12 | XP_016859245.1 | |
XM_017003757.1 | 454 | Missense Mutation | ACC,TCC | T,S 12 | XP_016859246.1 | |
XM_017003758.1 | 454 | Missense Mutation | ACC,TCC | T,S 12 | XP_016859247.1 |