Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAACGGACCCAGACCTCCCCGGT[A/G]GGGACCTCATGCAGCAGCAGTCGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604464 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITSN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ITSN2 - intersectin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006277.2 | 5211 | Silent Mutation | CCC,CCT | P,P 1678 | NP_006268.2 | |
NM_019595.3 | 5211 | Silent Mutation | CCC,CCT | P,P 1651 | NP_062541.3 | |
NM_147152.2 | 5211 | Intron | NP_671494.2 |