Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGCACCGGTCCATGAACCCCTGC[C/T]CTGTGCACGATGCTGGCACGGGCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608527 | ||||||||||||||||||||
Literature Links: |
LOC107986004 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC107986004 - uncharacterized LOC107986004 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NEU4 - neuraminidase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167599.2 | 979 | Missense Mutation | CCT,TCT | P,S 102 | NP_001161071.1 | |
NM_001167600.2 | 979 | Missense Mutation | CCT,TCT | P,S 89 | NP_001161072.1 | |
NM_001167601.2 | 979 | Missense Mutation | CCT,TCT | P,S 89 | NP_001161073.1 | |
NM_001167602.2 | 979 | Missense Mutation | CCT,TCT | P,S 89 | NP_001161074.1 | |
NM_080741.3 | 979 | Missense Mutation | CCT,TCT | P,S 101 | NP_542779.2 |