Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGATTTCTTTTTGTGAAACTTGT[A/G]TGATGCTGGCAGGTCATATCGAAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 109090 | ||||||||||||||||||||
Literature Links: |
METTL5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
METTL5 - methyltransferase like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001293186.1 | 761 | Missense Mutation | CAC,TAC | H,Y 190 | NP_001280115.1 | |
NM_001293187.1 | 761 | Missense Mutation | CAC,TAC | H,Y 190 | NP_001280116.1 | |
NM_014168.3 | 761 | Missense Mutation | CAC,TAC | H,Y 190 | NP_054887.2 | |
XM_011511023.1 | 761 | Silent Mutation | CAC,CAT | H,H 172 | XP_011509325.1 |
SSB - Sjogren syndrome antigen B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001294145.1 | 761 | Intron | NP_001281074.1 | |||
NM_003142.4 | 761 | Intron | NP_003133.1 |