Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTAGTCCATCCGAGCATCCTTTG[C/T]TGACAGTGCCATAGAGTTCCATTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607701 | ||||||||||||||||||||
Literature Links: |
FASTKD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FASTKD1 - FAST kinase domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281476.2 | 2802 | Missense Mutation | ACA,GCA | T,A 784 | NP_001268405.1 | |
NM_001322046.1 | 2802 | Missense Mutation | ACA,GCA | T,A 827 | NP_001308975.1 | |
NM_001322048.1 | 2802 | Missense Mutation | ACA,GCA | T,A 804 | NP_001308977.1 | |
NM_001322049.1 | 2802 | Missense Mutation | ACA,GCA | T,A 761 | NP_001308978.1 | |
NM_024622.5 | 2802 | Missense Mutation | ACA,GCA | T,A 827 | NP_078898.3 | |
XM_006712750.3 | 2802 | Missense Mutation | ACA,GCA | T,A 804 | XP_006712813.1 | |
XM_006712751.3 | 2802 | Missense Mutation | ACA,GCA | T,A 507 | XP_006712814.1 | |
XM_017004911.1 | 2802 | Missense Mutation | ACA,GCA | T,A 655 | XP_016860400.1 | |
XM_017004912.1 | 2802 | Missense Mutation | ACA,GCA | T,A 464 | XP_016860401.1 |
KLHL41 - kelch like family member 41 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |