Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTGTGCCTTTGTCACCAACCAGC[A/G]CACGCATGAACAGATGGGCCCCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PROM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PROM2 - prominin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165977.2 | 655 | Missense Mutation | CAC,CGC | H,R 178 | NP_001159449.1 | |
NM_001165978.2 | 655 | Missense Mutation | CAC,CGC | H,R 178 | NP_001159450.1 | |
NM_001321070.1 | 655 | UTR 5 | NP_001307999.1 | |||
NM_144707.3 | 655 | Missense Mutation | CAC,CGC | H,R 178 | NP_653308.2 | |
XM_006712301.2 | 655 | UTR 5 | XP_006712364.1 | |||
XM_011510672.2 | 655 | Missense Mutation | CAC,CGC | H,R 178 | XP_011508974.1 |