Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAACCTGTAAGGTCAGAGAGGGA[C/T]TCCTCTGACTTAGTCTTTCCCTTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606133 | ||||||||||||||||||||
Literature Links: |
CDC42EP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDC42EP3 - CDC42 effector protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270436.1 | 1675 | Silent Mutation | GAA,GAG | E,E 222 | NP_001257365.1 | |
NM_001270437.1 | 1675 | Silent Mutation | GAA,GAG | E,E 222 | NP_001257366.1 | |
NM_001270438.1 | 1675 | Silent Mutation | GAA,GAG | E,E 222 | NP_001257367.1 | |
NM_006449.4 | 1675 | Silent Mutation | GAA,GAG | E,E 222 | NP_006440.2 | |
XM_011532482.2 | 1675 | Silent Mutation | GAA,GAG | E,E 222 | XP_011530784.1 |