Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCATGGTCACTTTGTTATTCTTCA[A/G]GTGAACTTTGACAGTAAATCCTCGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607800 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCA12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCA12 - ATP binding cassette subfamily A member 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015657.3 | 7688 | Silent Mutation | CTG,TTG | L,L 2172 | NP_056472.2 | |
NM_173076.2 | 7688 | Silent Mutation | CTG,TTG | L,L 2490 | NP_775099.2 | |
XM_011510951.2 | 7688 | Silent Mutation | CTG,TTG | L,L 2493 | XP_011509253.1 |
LOC101928103 - uncharacterized LOC101928103 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |