Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTATACATCCAAAAAGAGGGAAT[A/G]ATCATGGCAATTAAAGCTGCCTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612032 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPY30 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DPY30 - dpy-30, histone methyltransferase complex regulatory subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321209.1 | 656 | UTR 3 | NP_001308138.1 | |||
NM_001321210.1 | 656 | Intron | NP_001308139.1 | |||
NM_032574.3 | 656 | UTR 3 | NP_115963.1 | |||
XM_006712117.3 | 656 | Intron | XP_006712180.1 | |||
XM_006712118.3 | 656 | Intron | XP_006712181.1 | |||
XM_011533133.2 | 656 | Intron | XP_011531435.1 | |||
XM_017005111.1 | 656 | Intron | XP_016860600.1 |