Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTAACACAGTAGCGAGGAAACACC[A/G]TTTGGGGATTGTCACTGTATGTTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616712 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
STARD7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STARD7 - StAR related lipid transfer domain containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020151.3 | 1288 | Missense Mutation | ACG,ATG | T,M 296 | NP_064536.2 | |
XM_011511499.1 | 1288 | Missense Mutation | ACG,ATG | T,M 195 | XP_011509801.1 |