Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCAACGTTGAAAATGTCCTTTGT[A/G]GGGGAGAACTCAGGAGTGAAAATGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605510 | ||||||||||||||||||||
Literature Links: |
IL37 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IL37 - interleukin 37 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014439.3 | 54 | Silent Mutation | GTA,GTG | V,V 4 | NP_055254.2 | |
NM_173202.1 | 54 | Silent Mutation | GTA,GTG | V,V 4 | NP_775294.1 | |
NM_173203.1 | 54 | Silent Mutation | GTA,GTG | V,V 4 | NP_775295.1 | |
NM_173204.1 | 54 | Silent Mutation | GTA,GTG | V,V 4 | NP_775296.1 | |
NM_173205.1 | 54 | Intron | NP_775297.1 | |||
XM_011510963.2 | 54 | Silent Mutation | GTA,GTG | V,V 4 | XP_011509265.1 | |
XM_011510964.2 | 54 | Silent Mutation | GTA,GTG | V,V 4 | XP_011509266.1 |