Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTGTTGTTGCAGGAAGTCCATCT[C/G]GCCATGCAATGGGCGCATCCTGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608058 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
G6PC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
G6PC2 - glucose-6-phosphatase catalytic subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081686.1 | 432 | Missense Mutation | CGC,GGC | R,G 114 | NP_001075155.1 | |
NM_021176.2 | 432 | Missense Mutation | CGC,GGC | R,G 114 | NP_066999.1 | |
XM_011511564.2 | 432 | Intron | XP_011509866.1 | |||
XM_011511565.2 | 432 | UTR 5 | XP_011509867.1 |