Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCAGCTCTTTTCAGGAGTAGACA[C/T]GCAATTGCCAAGGAATTTCATGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 171890 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDE1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDE1A - phosphodiesterase 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003683.2 | Intron | NP_001003683.1 | ||||
NM_001258312.1 | Intron | NP_001245241.1 | ||||
NM_001258313.1 | Intron | NP_001245242.1 | ||||
NM_001258314.1 | Intron | NP_001245243.1 | ||||
NM_005019.4 | Intron | NP_005010.2 | ||||
XM_011511323.2 | Intron | XP_011509625.1 | ||||
XM_011511324.2 | Intron | XP_011509626.1 | ||||
XM_011511325.2 | Intron | XP_011509627.1 | ||||
XM_011511326.2 | Intron | XP_011509628.1 | ||||
XM_017004294.1 | Intron | XP_016859783.1 | ||||
XM_017004295.1 | Intron | XP_016859784.1 | ||||
XM_017004296.1 | Intron | XP_016859785.1 | ||||
XM_017004297.1 | Intron | XP_016859786.1 | ||||
XM_017004298.1 | Intron | XP_016859787.1 | ||||
XM_017004299.1 | Intron | XP_016859788.1 | ||||
XM_017004300.1 | Intron | XP_016859789.1 | ||||
XM_017004301.1 | Intron | XP_016859790.1 | ||||
XM_017004302.1 | Intron | XP_016859791.1 |