Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATAACTTGATATTTTATATAAACA[C/T]TGGCCTCTACTAACAGCCTTTTCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607871 MIM: 600678 | ||||||||||||||||||||
Literature Links: |
FBXO11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBXO11 - F-box protein 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190274.1 | 2522 | Silent Mutation | CAA,CAG | Q,Q 834 | NP_001177203.1 | |
NM_025133.4 | 2522 | Silent Mutation | CAA,CAG | Q,Q 750 | NP_079409.3 | |
XM_005264572.4 | 2522 | Silent Mutation | CAA,CAG | Q,Q 834 | XP_005264629.1 | |
XM_005264573.4 | 2522 | Silent Mutation | CAA,CAG | Q,Q 833 | XP_005264630.1 | |
XM_017005015.1 | 2522 | Silent Mutation | CAA,CAG | Q,Q 833 | XP_016860504.1 | |
XM_017005016.1 | 2522 | Silent Mutation | CAA,CAG | Q,Q 750 | XP_016860505.1 | |
XM_017005017.1 | 2522 | Silent Mutation | CAA,CAG | Q,Q 750 | XP_016860506.1 |
MSH6 - mutS homolog 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |