Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAATCATCACAGGTTCCAGCAGT[C/T]GGAGCAGACCCTGGAGCACCAGTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ATP6V1E2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATP6V1E2 - ATPase H+ transporting V1 subunit E2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318063.1 | 1822 | Missense Mutation | NP_001304992.1 | |||
XM_005264633.2 | 1822 | Missense Mutation | XP_005264690.1 | |||
XM_005264634.2 | 1822 | Missense Mutation | XP_005264691.1 | |||
XM_011533148.2 | 1822 | Missense Mutation | XP_011531450.1 | |||
XM_011533149.2 | 1822 | Missense Mutation | XP_011531451.1 | |||
XM_011533150.2 | 1822 | Missense Mutation | XP_011531452.1 | |||
XM_011533151.2 | 1822 | Missense Mutation | XP_011531453.1 | |||
XM_011533152.2 | 1822 | Missense Mutation | XP_011531454.1 | |||
XM_011533153.2 | 1822 | Missense Mutation | XP_011531455.1 | |||
XM_017005225.1 | 1822 | Missense Mutation | XP_016860714.1 | |||
XM_017005226.1 | 1822 | Missense Mutation | XP_016860715.1 |