Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTTTTTCTCTTATGCTTAGACC[A/G]CTCCTCCTCTGATTTTTCTCTGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
KRCC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KRCC1 - lysine rich coiled-coil 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304526.1 | 1290 | Missense Mutation | CGG,TGG | R,W 175 | NP_001291455.1 | |
NM_016618.2 | 1290 | Missense Mutation | CGG,TGG | R,W 175 | NP_057702.1 | |
XM_017004292.1 | 1290 | Missense Mutation | CGG,TGG | R,W 175 | XP_016859781.1 |