Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGTCTTGGCTTACTTTACACAAA[A/G]CAAACTGGCCCAATTTCAACGCCGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 120190 | ||||||||||||||||||||
Literature Links: |
COL5A2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COL5A2 - collagen type V alpha 2 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000393.3 | 4548 | Silent Mutation | TGC,TGT | C,C 1497 | NP_000384.2 | |
XM_011510573.2 | 4548 | Silent Mutation | TGC,TGT | C,C 1451 | XP_011508875.1 |