Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGCCAGGAGTACTTCCACCCCGG[C/T]GGGGGCAGCCCGGCCGCTGCCTACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142980 MIM: 142985 MIM: 142982 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HOXD-AS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HOXD-AS2 - HOXD cluster antisense RNA 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXD3 - homeobox D3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXD8 - homeobox D8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199746.1 | 918 | Silent Mutation | GGC,GGT | G,G 97 | NP_001186675.1 | |
NM_001199747.1 | 918 | Intron | NP_001186676.1 | |||
NM_019558.3 | 918 | Silent Mutation | GGC,GGT | G,G 97 | NP_062458.1 |
HOXD9 - homeobox D9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100129455 - uncharacterized LOC100129455 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |