Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATTCAAGCACTTTTTAATTCCAG[G/T]TTCATCTGTGTGACTGTCCTTGGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611751 | ||||||||||||||||||||
Literature Links: |
THUMPD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
THUMPD2 - THUMP domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321468.1 | 1441 | UTR 3 | NP_001308397.1 | |||
NM_001321469.1 | 1441 | Missense Mutation | NP_001308398.1 | |||
NM_001321470.1 | 1441 | Missense Mutation | NP_001308399.1 | |||
NM_001321474.1 | 1441 | UTR 3 | NP_001308403.1 | |||
NM_001321475.1 | 1441 | UTR 3 | NP_001308404.1 | |||
NM_001321477.1 | 1441 | Missense Mutation | NP_001308406.1 | |||
NM_001321478.1 | 1441 | Missense Mutation | NP_001308407.1 | |||
NM_001321479.1 | 1441 | Missense Mutation | NP_001308408.1 | |||
NM_001321480.1 | 1441 | Missense Mutation | NP_001308409.1 | |||
NM_001321481.1 | 1441 | Missense Mutation | NP_001308410.1 | |||
NM_025264.4 | 1441 | Missense Mutation | NP_079540.2 | |||
XM_011533119.2 | 1441 | Missense Mutation | XP_011531421.1 | |||
XM_017005050.1 | 1441 | Missense Mutation | XP_016860539.1 | |||
XM_017005051.1 | 1441 | UTR 3 | XP_016860540.1 | |||
XM_017005052.1 | 1441 | Intron | XP_016860541.1 | |||
XM_017005053.1 | 1441 | Intron | XP_016860542.1 | |||
XM_017005054.1 | 1441 | Missense Mutation | XP_016860543.1 |