Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTCATTCCTCAGCTGCTCACCTG[A/G]TACCTCATACGAGTGAGTATAGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605843 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PECR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PECR - peroxisomal trans-2-enoyl-CoA reductase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018441.5 | 898 | Missense Mutation | CCA,TCA | P,S 275 | NP_060911.2 | |
XM_011511478.2 | 898 | Missense Mutation | CCA,TCA | P,S 129 | XP_011509780.1 |