Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTCAAGTGTATTTCTGTGTATG[A/G]AGAAACAGTAACTCTATGGCAGGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603525 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NMI PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NMI - N-myc and STAT interactor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004688.2 | 1112 | Missense Mutation | CCA,TCA | P,S 237 | NP_004679.2 | |
XM_005246941.2 | 1112 | Missense Mutation | CCA,TCA | P,S 237 | XP_005246998.1 | |
XM_017005247.1 | 1112 | Missense Mutation | CCA,TCA | P,S 237 | XP_016860736.1 |
RBM43 - RNA binding motif protein 43 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |