Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGCCGCGGCAGGATGATTGCCTC[A/G]CATCTGCTTGCCTACTTCTTCACGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601125 | ||||||||||||||||||||
Literature Links: |
HK2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HK2 - hexokinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000189.4 | 1938 | Silent Mutation | TCA,TCG | S,S 4 | NP_000180.2 | |
XM_005264280.1 | 1938 | Intron | XP_005264337.1 | |||
XM_011532807.1 | 1938 | Intron | XP_011531109.1 | |||
XM_017003945.1 | 1938 | Intron | XP_016859434.1 |