Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTCCCCTTACTGGAAAAGGTCTA[C/T]GCCAAGTGCGTGTGCTGGGGGCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605286 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPN10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAPN10 - calpain 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023083.3 | 661 | Silent Mutation | TAC,TAT | Y,Y 155 | NP_075571.1 | |
NM_023085.3 | 661 | Silent Mutation | TAC,TAT | Y,Y 155 | NP_075573.2 |
CAPN10-AS1 - CAPN10 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |