Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCCTCCTACAACCTGTTGAGTGC[A/G]TATACTTGGCCAACTTCTGTATAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613596 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM161A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM161A - family with sequence similarity 161 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201543.1 | Intron | NP_001188472.1 | ||||
NM_032180.2 | Intron | NP_115556.2 | ||||
XM_017005072.1 | Intron | XP_016860561.1 | ||||
XM_017005073.1 | Intron | XP_016860562.1 | ||||
XM_017005074.1 | Intron | XP_016860563.1 |