Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAATGTATTTCACCAGTAGCAAA[C/T]GAGATGAACCATCTTCCTGCACACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604585 | ||||||||||||||||||||
Literature Links: |
SP100 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SP100 - SP100 nuclear antigen | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080391.1 | 160 | Silent Mutation | AAC,AAT | N,N 22 | NP_001073860.1 | |
NM_001206701.1 | 160 | Silent Mutation | AAC,AAT | N,N 22 | NP_001193630.1 | |
NM_001206702.1 | 160 | Silent Mutation | AAC,AAT | N,N 22 | NP_001193631.1 | |
NM_001206703.1 | 160 | Intron | NP_001193632.1 | |||
NM_001206704.1 | 160 | UTR 5 | NP_001193633.1 | |||
NM_003113.3 | 160 | Silent Mutation | AAC,AAT | N,N 22 | NP_003104.2 |