Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCGATCACAGGCAGTAGCAAAC[A/G]AGAGGGCACACCCTGCCAGCACAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615129 | ||||||||||||||||||||
Literature Links: |
GALNT5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GALNT5 - polypeptide N-acetylgalactosaminyltransferase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017003237.1 | 1244 | Missense Mutation | AAG,GAG | K,E 237 | XP_016858726.1 | |
XM_017003238.1 | 1244 | Missense Mutation | AAG,GAG | K,E 237 | XP_016858727.1 |