Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGATGCAAACAAGCATGAAGACTC[C/T]GAGGAGTATGTGGTCCATCACCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 100690 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHRNA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHRNA1 - cholinergic receptor nicotinic alpha 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000079.3 | 1372 | Missense Mutation | AGA,GGA | R,G 432 | NP_000070.1 | |
NM_001039523.2 | 1372 | Missense Mutation | AGA,GGA | R,G 457 | NP_001034612.1 | |
XM_017003256.1 | 1372 | Missense Mutation | AGA,GGA | R,G 464 | XP_016858745.1 | |
XM_017003257.1 | 1372 | Missense Mutation | AGA,GGA | R,G 439 | XP_016858746.1 |