Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTGCCCTGTATTTTGCTTCTGTC[C/T]GTGAATTCTGTTGCTTCTGGGGCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605734 | ||||||||||||||||||||
Literature Links: |
TMEFF2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEFF2 - transmembrane protein with EGF like and two follistatin like domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305134.1 | 1503 | UTR 3 | NP_001292063.1 | |||
NM_001305145.1 | 1503 | Intron | NP_001292074.1 | |||
NM_016192.3 | 1503 | Missense Mutation | AGA,GGA | R,G 353 | NP_057276.2 | |
XM_011510890.2 | 1503 | Missense Mutation | AGA,GGA | R,G 344 | XP_011509192.1 | |
XM_017003739.1 | 1503 | UTR 3 | XP_016859228.1 | |||
XM_017003740.1 | 1503 | Intron | XP_016859229.1 |