Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAGACACCAGCTCATTCCTCTGG[A/G]CAGCCTCCCAGGGATGCTAAGTTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606622 | ||||||||||||||||||||
Literature Links: |
SMARCAL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SMARCAL1 - SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127207.1 | 690 | Silent Mutation | GGA,GGG | G,G 182 | NP_001120679.1 | |
NM_014140.3 | 690 | Silent Mutation | GGA,GGG | G,G 182 | NP_054859.2 | |
XM_005246631.2 | 690 | Silent Mutation | GGA,GGG | G,G 182 | XP_005246688.1 | |
XM_005246632.1 | 690 | Silent Mutation | GGA,GGG | G,G 182 | XP_005246689.1 | |
XM_017004228.1 | 690 | UTR 5 | XP_016859717.1 |