Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTGGGAAGATGGCAGGTGGGGGC[A/G]GCGACCTGAGCACCAGGTGAGTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604585 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SP100 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SP100 - SP100 nuclear antigen | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080391.1 | 157 | Missense Mutation | AGC,GGC | S,G 6 | NP_001073860.1 | |
NM_001206701.1 | 157 | Missense Mutation | AGC,GGC | S,G 6 | NP_001193630.1 | |
NM_001206702.1 | 157 | Missense Mutation | AGC,GGC | S,G 6 | NP_001193631.1 | |
NM_001206703.1 | 157 | Missense Mutation | AGC,GGC | S,G 6 | NP_001193632.1 | |
NM_001206704.1 | 157 | Intron | NP_001193633.1 | |||
NM_003113.3 | 157 | Missense Mutation | AGC,GGC | S,G 6 | NP_003104.2 |
SP140L - SP140 nuclear body protein like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |