Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGCGCCCGCGCGCACTTCAAGGA[A/G]CTGGTGAACGCCGTGGCCACTGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601181 MIM: 611737 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RANBP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RANBP2 - RAN binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006267.4 | 330 | Intron | NP_006258.3 | |||
XM_005264002.2 | 330 | Intron | XP_005264059.1 | |||
XM_005264003.2 | 330 | Intron | XP_005264060.1 | |||
XM_005264004.2 | 330 | Intron | XP_005264061.1 | |||
XM_005264005.4 | 330 | Intron | XP_005264062.1 | |||
XM_005264007.2 | 330 | Intron | XP_005264064.1 | |||
XM_011511575.2 | 330 | Intron | XP_011509877.1 | |||
XM_011511576.2 | 330 | Intron | XP_011509878.1 | |||
XM_011511578.2 | 330 | Intron | XP_011509880.1 | |||
XM_017004623.1 | 330 | Intron | XP_016860112.1 | |||
XM_017004624.1 | 330 | Intron | XP_016860113.1 | |||
XM_017004625.1 | 330 | Intron | XP_016860114.1 |
SOWAHC - sosondowah ankyrin repeat domain family member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023016.3 | 330 | Silent Mutation | GAA,GAG | E,E 58 | NP_075392.2 |