Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCTTTTCAGGTTGGCTCTGACA[C/T]GTTTTATTCCTTCAAGTACGAAATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612358 | ||||||||||||||||||||
Literature Links: |
KNG1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KNG1 - kininogen 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000893.3 | 443 | Missense Mutation | ACG,ATG | T,M 70 | NP_000884.1 | |
NM_001102416.2 | 443 | Missense Mutation | ACG,ATG | T,M 70 | NP_001095886.1 | |
NM_001166451.1 | 443 | Missense Mutation | ACG,ATG | T,M 70 | NP_001159923.1 |