Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGGTCTTCACCTTGTTCTGTTG[C/T]GTATTTTTGTTTGTGAATTACCTTG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 616933 MIM: 613516 | |||||||||||||||||||||||
Literature Links: |
FYTTD1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
FYTTD1 - forty-two-three domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011537.2 | Intron | NP_001011537.2 | ||||
NM_032288.6 | Intron | NP_115664.2 |
RUBCN - RUN and cysteine rich domain containing beclin 1 interacting protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |