Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATTTAGAGAAGACTTTTCAGGTC[C/T]GTCGACCTGCGGGCACTATCTTACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609380 | ||||||||||||||||||||
Literature Links: |
LMLN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LMLN - leishmanolysin like peptidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136049.2 | 405 | Missense Mutation | CGT,TGT | R,C 134 | NP_001129521.2 | |
NM_033029.3 | 405 | Missense Mutation | CGT,TGT | R,C 134 | NP_149018.2 | |
XM_011513270.1 | 405 | Missense Mutation | CGT,TGT | R,C 134 | XP_011511572.1 | |
XM_011513271.1 | 405 | Missense Mutation | CGT,TGT | R,C 134 | XP_011511573.1 | |
XM_017007428.1 | 405 | Missense Mutation | CGT,TGT | R,C 134 | XP_016862917.1 |