Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCTTTGAGAACCTGTCCAGGGG[C/T]CTGGGTATGGCCTACTTCCCAGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 142408 MIM: 614472 | ||||||||||||||||||||
Literature Links: |
MST1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MST1 - macrophage stimulating 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF123 - ring finger protein 123 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022064.4 | 828 | Silent Mutation | GGC,GGT | G,G 228 | NP_071347.2 | |
XM_006713285.1 | 828 | Silent Mutation | GGC,GGT | G,G 228 | XP_006713348.1 | |
XM_011533995.1 | 828 | Silent Mutation | GGC,GGT | G,G 228 | XP_011532297.1 | |
XM_017007018.1 | 828 | Silent Mutation | GGC,GGT | G,G 228 | XP_016862507.1 |