Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTGGTCCAGCACGTCTTGGTGGT[A/G]CACGAGCCTGACACGCCGTGGCACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601570 | ||||||||||||||||||||
Literature Links: |
WNT7A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WNT7A - Wnt family member 7A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004625.3 | 932 | Silent Mutation | TGC,TGT | C,C 209 | NP_004616.2 | |
XM_011534091.2 | 932 | Silent Mutation | TGC,TGT | C,C 142 | XP_011532393.1 |