Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACCAACAAGAATGAGGAGCAGGA[C/G]ATCTTCATTTACAGAGCAACTTGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602509 | ||||||||||||||||||||
Literature Links: |
GOLGA4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GOLGA4 - golgin A4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172713.1 | 476 | Missense Mutation | ACA,AGA | T,R 39 | NP_001166184.1 | |
NM_002078.4 | 476 | Missense Mutation | ACA,AGA | T,R 39 | NP_002069.2 | |
XM_005265069.3 | 476 | Missense Mutation | ACA,AGA | T,R 39 | XP_005265126.1 | |
XM_005265070.3 | 476 | Missense Mutation | ACA,AGA | T,R 39 | XP_005265127.1 | |
XM_005265071.3 | 476 | Missense Mutation | ACA,AGA | T,R 39 | XP_005265128.1 | |
XM_005265072.3 | 476 | Missense Mutation | ACA,AGA | T,R 39 | XP_005265129.1 | |
XM_005265073.1 | 476 | Missense Mutation | ACA,AGA | T,R 39 | XP_005265130.1 | |
XM_005265074.1 | 476 | Missense Mutation | ACA,AGA | T,R 39 | XP_005265131.1 | |
XM_005265075.1 | 476 | Missense Mutation | ACA,AGA | T,R 39 | XP_005265132.1 | |
XM_006713110.2 | 476 | Missense Mutation | ACA,AGA | T,R 39 | XP_006713173.1 | |
XM_011533598.2 | 476 | Intron | XP_011531900.1 | |||
XM_017006184.1 | 476 | Missense Mutation | ACA,AGA | T,R 39 | XP_016861673.1 | |
XM_017006185.1 | 476 | Missense Mutation | ACA,AGA | T,R 5 | XP_016861674.1 | |
XM_017006186.1 | 476 | Missense Mutation | ACA,AGA | T,R 5 | XP_016861675.1 | |
XM_017006187.1 | 476 | Missense Mutation | ACA,AGA | T,R 39 | XP_016861676.1 | |
XM_017006188.1 | 476 | Missense Mutation | ACA,AGA | T,R 39 | XP_016861677.1 |
LOC152048 - uncharacterized FLJ31715 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |